missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-4,6-dichloropyrimidine 98.0+%, TCI America™

Catalog No. A13951G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13951G 1 g
A13955G 5 g
A139525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A13951G Supplier TCI America Supplier No. A13951G

Chemical Identifiers

CAS 5413-85-4
Molecular Formula C4H3Cl2N3
Molecular Weight (g/mol) 163.99
MDL Number MFCD00006108
InChI Key NIGDWBHWHVHOAD-UHFFFAOYSA-N
Synonym 5-amino-4,6-dichloropyrimidine, 5-pyrimidinamine, 4,6-dichloro, 4,6-dichloro-5-pyrimidinamine, 4,6-dichloro-5-aminopyrimidine, 4,6-dichloropyrimidin-5-ylamine, 5-amine-4,6-dichloropyrimidine, 5-amino-4,6-dichloropyfimidine, 4,6-dichloro-pyrimidin-5-ylamine, pyrimidine, 5-amino-4,6-dichloro, 4,6-dichloropyrimidine-5-ylamine
PubChem CID 79434
IUPAC Name 4,6-dichloropyrimidin-5-amine
SMILES NC1=C(Cl)N=CN=C1Cl

Specifications

Melting Point 144°C
Color White-Yellow
Quantity 1 g
Formula Weight 163.99
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-4,6-dichloropyrimidine