missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-4,6-dichloropyrimidine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 5413-85-4
Molecular Formula C4H3Cl2N3
Molecular Weight (g/mol) 163.99
MDL Number MFCD00006108
InChI Key NIGDWBHWHVHOAD-UHFFFAOYSA-N
Synonym 5-amino-4,6-dichloropyrimidine, 5-pyrimidinamine, 4,6-dichloro, 4,6-dichloro-5-pyrimidinamine, 4,6-dichloro-5-aminopyrimidine, 4,6-dichloropyrimidin-5-ylamine, 5-amine-4,6-dichloropyrimidine, 5-amino-4,6-dichloropyfimidine, 4,6-dichloro-pyrimidin-5-ylamine, pyrimidine, 5-amino-4,6-dichloro, 4,6-dichloropyrimidine-5-ylamine
PubChem CID 79434
IUPAC Name 4,6-dichloropyrimidin-5-amine
SMILES NC1=C(Cl)N=CN=C1Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC203250050
SDS View Documents
Thermo Scientific Chemicals
203250050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC203250250
SDS View Documents
Thermo Scientific Chemicals
203250250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

5413-85-4
163.99
NIGDWBHWHVHOAD-UHFFFAOYSA-N
79434
NC1=C(Cl)N=CN=C1Cl
C4H3Cl2N3
MFCD00006108
5-amino-4,6-dichloropyrimidine, 5-pyrimidinamine, 4,6-dichloro, 4,6-dichloro-5-pyrimidinamine, 4,6-dichloro-5-aminopyrimidine, 4,6-dichloropyrimidin-5-ylamine, 5-amine-4,6-dichloropyrimidine, 5-amino-4,6-dichloropyfimidine, 4,6-dichloro-pyrimidin-5-ylamine, pyrimidine, 5-amino-4,6-dichloro, 4,6-dichloropyrimidine-5-ylamine
4,6-dichloropyrimidin-5-amine

Specifications

5413-85-4
100.0
96% min. (HPLC)
C4H3Cl2N3
5g
Solubility in water: slightly soluble
NC1=C(Cl)N=CN=C1Cl
163.99
163.99
5-Amino-4, 6-dichloropyrimidine
96.0
Authentic
Glass bottle
MFCD00006108
5-amino-4,6-dichloropyrimidine, 5-pyrimidinamine, 4,6-dichloro, 4,6-dichloro-5-pyrimidinamine, 4,6-dichloro-5-aminopyrimidine, 4,6-dichloropyrimidin-5-ylamine, 5-amine-4,6-dichloropyrimidine, 5-amino-4,6-dichloropyfimidine, 4,6-dichloro-pyrimidin-5-ylamine, pyrimidine, 5-amino-4,6-dichloro, 4,6-dichloropyrimidine-5-ylamine
NIGDWBHWHVHOAD-UHFFFAOYSA-N
4,6-dichloropyrimidin-5-amine
79434
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 226-503-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only