missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 39906-04-2 |
|---|---|
| Molecular Formula | C5H5Cl2N3 |
| Molecular Weight (g/mol) | 178.02 |
| InChI Key | FKRXXAMAHOGYNT-UHFFFAOYSA-N |
| Synonym | 5-amino-4,6-dichloro-2-methylpyrimidine, 2-methyl-4,6-dichloro-5-aminopyrimidine, 4,6-dichloro-2-methyl-pyrimidin-5-ylamine, 5-pyrimidinamine, 4,6-dichloro-2-methyl, 4,6-dichloro-2-methyl-pyrimidin-5-amine, 4,6-dichloro-2-methylpyrimidin-5-ylamine, pubchem6960, acmc-209j8x, ksc496o2d, fkrxxamahogynt-uhfffaoysa |
| PubChem CID | 736674 |
| IUPAC Name | 4,6-dichloro-2-methylpyrimidin-5-amine |
| SMILES | CC1=NC(=C(C(=N1)Cl)N)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC442520010
|
N/A | 1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 39906-04-2 | |
| 178.02 | |
| 5-amino-4,6-dichloro-2-methylpyrimidine, 2-methyl-4,6-dichloro-5-aminopyrimidine, 4,6-dichloro-2-methyl-pyrimidin-5-ylamine, 5-pyrimidinamine, 4,6-dichloro-2-methyl, 4,6-dichloro-2-methyl-pyrimidin-5-amine, 4,6-dichloro-2-methylpyrimidin-5-ylamine, pubchem6960, acmc-209j8x, ksc496o2d, fkrxxamahogynt-uhfffaoysa | |
| 4,6-dichloro-2-methylpyrimidin-5-amine |
| C5H5Cl2N3 | |
| FKRXXAMAHOGYNT-UHFFFAOYSA-N | |
| 736674 | |
| CC1=NC(=C(C(=N1)Cl)N)Cl |
Specifications
| 39906-04-2 | |
| Glass Bottle | |
| 5-amino-4,6-dichloro-2-methylpyrimidine, 2-methyl-4,6-dichloro-5-aminopyrimidine, 4,6-dichloro-2-methyl-pyrimidin-5-ylamine, 5-pyrimidinamine, 4,6-dichloro-2-methyl, 4,6-dichloro-2-methyl-pyrimidin-5-amine, 4,6-dichloro-2-methylpyrimidin-5-ylamine, pubchem6960, acmc-209j8x, ksc496o2d, fkrxxamahogynt-uhfffaoysa | |
| CC1=NC(=C(C(=N1)Cl)N)Cl | |
| 178.02 | |
| 178.02 | |
| 5-Amino-4, 6-dichloro-2-methylpyrimidine |
| 96% min. (HPLC) | |
| C5H5Cl2N3 | |
| FKRXXAMAHOGYNT-UHFFFAOYSA-N | |
| 4,6-dichloro-2-methylpyrimidin-5-amine | |
| 736674 | |
| 97% |
Safety and Handling
GHS Signal Word: Warning
RUO – Research Use Only