missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-3-methylisoxazole 97.0+%, TCI America™

Catalog No. p-7118120
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28671G 1 g
A28675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28671G Supplier TCI America Supplier No. A28671G
Only null left

Chemical Identifiers

CAS 14678-02-5
Molecular Formula C4H6N2O
Molecular Weight (g/mol) 98.105
MDL Number MFCD00003151
InChI Key FNXYWHTZDAVRTB-UHFFFAOYSA-N
Synonym 5-amino-3-methylisoxazole, 3-methylisoxazol-5-amine, 5-isoxazolamine, 3-methyl, 3-methyl-5-isoxazolamine, unii-0g66r1h4rw, 3-methyl-isoxazol-5-ylamine, 3-methylisoxazole-5-ylamine, 3-methylisoxazole-5-amine, acmc-209yk8, 3-methylisooxazole-5-amine
PubChem CID 84590
IUPAC Name 3-methyl-1,2-oxazol-5-amine
SMILES CC1=NOC(=C1)N

Specifications

Melting Point 84°C
Quantity 1 g
Formula Weight 98.11
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-3-methylisoxazole