missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-3-methylisoxazole, 98%

Catalog No. AAA1244603
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1244603 1 g
AAA1244606 5 g
AAA1244614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1244603 Supplier Thermo Scientific Chemicals Supplier No. A1244603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 14678-02-5 | C4H6N2O | 98.105 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14678-02-5
Molecular Formula C4H6N2O
Molecular Weight (g/mol) 98.105
MDL Number MFCD00003151
InChI Key FNXYWHTZDAVRTB-UHFFFAOYSA-N
Synonym 5-amino-3-methylisoxazole, 3-methylisoxazol-5-amine, 5-isoxazolamine, 3-methyl, 3-methyl-5-isoxazolamine, unii-0g66r1h4rw, 3-methyl-isoxazol-5-ylamine, 3-methylisoxazole-5-ylamine, 3-methylisoxazole-5-amine, acmc-209yk8, 3-methylisooxazole-5-amine
PubChem CID 84590
IUPAC Name 3-methyl-1,2-oxazol-5-amine
SMILES CC1=NOC(=C1)N

Specifications

Melting Point 81°C to 85°C
Quantity 1 g
Beilstein 107906
Formula Weight 98.11
Percent Purity ≥98%
Chemical Name or Material 5-Amino-3-methylisoxazole
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 238-719-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only