missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-3-methyl-1-p-tolylpyrazole 97.0+%, TCI America™
$242.33 - $242.33
Chemical Identifiers
| CAS | 62535-60-8 |
|---|---|
| Molecular Formula | C11H13N3 |
| Molecular Weight (g/mol) | 187.246 |
| MDL Number | MFCD00020725 |
| InChI Key | WQUNBEPKWJLYJD-UHFFFAOYSA-N |
| PubChem CID | 112855 |
| IUPAC Name | 5-methyl-2-(4-methylphenyl)pyrazol-3-amine |
| SMILES | CC1=CC=C(C=C1)N2C(=CC(=N2)C)N |
Chemical Identifiers
| 62535-60-8 | |
| 187.246 | |
| WQUNBEPKWJLYJD-UHFFFAOYSA-N | |
| 5-methyl-2-(4-methylphenyl)pyrazol-3-amine |
| C11H13N3 | |
| MFCD00020725 | |
| 112855 | |
| CC1=CC=C(C=C1)N2C(=CC(=N2)C)N |
Specifications
| 62535-60-8 | |
| C11H13N3 | |
| 10 g | |
| CC1=CC=C(C=C1)N2C(=CC(=N2)C)N | |
| 187.246 | |
| 187.25 | |
| Crystalline Powder |
| 118°C | |
| MFCD00020725 | |
| WQUNBEPKWJLYJD-UHFFFAOYSA-N | |
| 5-methyl-2-(4-methylphenyl)pyrazol-3-amine | |
| 112855 | |
| ≥97.0% (GC,T) | |
| 5-Amino-3-methyl-1-p-tolylpyrazole |
Safety and Handling
TSCA : Yes