accessibility menu, dialog, popup

UserName

CH3NNCH3H2N

CAS RN 62535-60-8

IUPAC Name: 3-methyl-1-(4-methylphenyl)-1H-pyrazol-5-amine
Synonyms: 5-methyl-2-(4-methylphenyl)pyrazol-3-amine
Molecular Weight (g/mol): 187.25
Molecular Formula: C11H13N3
InChi Key: WQUNBEPKWJLYJD-UHFFFAOYSA-N
SMILES: CC1=NN(C(N)=C1)C1=CC=C(C)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results