missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-3-bromo-2-methylpyridine 98.0+%, TCI America™

Catalog No. p-7118186
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29961G 1 g
A29965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29961G Supplier TCI America Supplier No. A29961G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 186593-43-1 | C6H7BrN2 | 187.04 g/mol

Chemical Identifiers

CAS 186593-43-1
Molecular Formula C6H7BrN2
Molecular Weight (g/mol) 187.04
MDL Number MFCD08060462
InChI Key UCLLYIVDXZGQSZ-UHFFFAOYSA-N
Synonym 5-Amino-3-bromo-2-picoline
PubChem CID 18458896
IUPAC Name 5-bromo-6-methylpyridin-3-amine
SMILES CC1=C(C=C(C=N1)N)Br

Specifications

Melting Point 106°C
Quantity 1 g
Formula Weight 187.04
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-3-bromo-2-methylpyridine