accessibility menu, dialog, popup

UserName

CAS RN 186593-43-1

IUPAC Name: 5-bromo-6-methylpyridin-3-amine
Synonyms: 5-bromo-6-methylpyridin-3-amine
Molecular Weight (g/mol): 187.04
Molecular Formula: C6H7BrN2
InChi Key: UCLLYIVDXZGQSZ-UHFFFAOYSA-N
SMILES: CC1=C(Br)C=C(N)C=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results