missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-3-(4-methoxyphenyl)-1-phenylpyrazole 98.0+%, TCI America™

Catalog No. p-7118204
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30551G 1 g
A30555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A30551G Supplier TCI America Supplier No. A30551G

Chemical Identifiers

CAS 19652-13-2
Molecular Formula C16H15N3O
Molecular Weight (g/mol) 265.32
MDL Number MFCD00197061
InChI Key LMLGFNHDEGGHOJ-UHFFFAOYSA-N
PubChem CID 4056290
IUPAC Name 3-(4-methoxyphenyl)-1-phenyl-1H-pyrazol-5-amine
SMILES COC1=CC=C(C=C1)C1=NN(C(N)=C1)C1=CC=CC=C1

Specifications

Melting Point 190°C
Quantity 1 g
Formula Weight 265.32
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-3-(4-methoxyphenyl)-1-phenylpyrazole