missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-3-(2-thienyl)-1H-pyrazole, 98%

CAS: 96799-03-0 | C7H7N3S | 165.21 g/mol

$70.78 - $241.85

Chemical Identifiers

CAS 96799-03-0
Molecular Formula C7H7N3S
Molecular Weight (g/mol) 165.21
MDL Number MFCD00051815
InChI Key TXSOLYKLZBJHFF-UHFFFAOYSA-N
Synonym 5-amino-3-2-thienyl pyrazole, 3-thiophen-2-yl-1h-pyrazol-5-amine, 5-thien-2-yl-1h-pyrazol-3-amine, 3-2-thienyl-1h-pyrazol-5-amine, 5-2-thienyl-1h-pyrazol-3-amine, 5-thiophen-2-yl-2h-pyrazol-3-ylamine, 5-thiophen-2-yl-1h-pyrazol-3-amine, gnf-pf-1850, 5-amino-3-thien-2-yl pyrazole, 5-amino-3-2-thienyl-1h-pyrazole
PubChem CID 523184
IUPAC Name 5-thiophen-2-yl-1H-pyrazol-3-amine
SMILES NC1=NNC(=C1)C1=CC=CS1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0675203
View Documents
Thermo Scientific Chemicals
L0675203
1 g
Each for $70.78
Only null left
 
AAL0675206
View Documents
Thermo Scientific Chemicals
L0675206
5 g
Each for $241.85
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

96799-03-0
165.21
TXSOLYKLZBJHFF-UHFFFAOYSA-N
523184
NC1=NNC(=C1)C1=CC=CS1
C7H7N3S
MFCD00051815
5-amino-3-2-thienyl pyrazole, 3-thiophen-2-yl-1h-pyrazol-5-amine, 5-thien-2-yl-1h-pyrazol-3-amine, 3-2-thienyl-1h-pyrazol-5-amine, 5-2-thienyl-1h-pyrazol-3-amine, 5-thiophen-2-yl-2h-pyrazol-3-ylamine, 5-thiophen-2-yl-1h-pyrazol-3-amine, gnf-pf-1850, 5-amino-3-thien-2-yl pyrazole, 5-amino-3-2-thienyl-1h-pyrazole
5-thiophen-2-yl-1H-pyrazol-3-amine

Specifications

96799-03-0
C7H7N3S
1 g
5-amino-3-2-thienyl pyrazole, 3-thiophen-2-yl-1h-pyrazol-5-amine, 5-thien-2-yl-1h-pyrazol-3-amine, 3-2-thienyl-1h-pyrazol-5-amine, 5-2-thienyl-1h-pyrazol-3-amine, 5-thiophen-2-yl-2h-pyrazol-3-ylamine, 5-thiophen-2-yl-1h-pyrazol-3-amine, gnf-pf-1850, 5-amino-3-thien-2-yl pyrazole, 5-amino-3-2-thienyl-1h-pyrazole
NC1=NNC(=C1)C1=CC=CS1
165.21
165.21
5-Amino-3-(2-thienyl)-1H-pyrazole
101°C to 103°C
MFCD00051815
4399374
TXSOLYKLZBJHFF-UHFFFAOYSA-N
5-thiophen-2-yl-1H-pyrazol-3-amine
523184
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H302-H315-H318-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only