missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-3-(2-thienyl)-1H-pyrazole, 98%

CAS: 96799-03-0 | C7H7N3S | 165.21 g/mol

Supplier:  Thermo Scientific Chemicals L0675206

 View more versions of this product

Catalog No. AAL0675206


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Amino-3-(2-thienyl)-1H-pyrazole
101°C to 103°C
MFCD00051815
4399374
TXSOLYKLZBJHFF-UHFFFAOYSA-N
5-thiophen-2-yl-1H-pyrazol-3-amine
523184
98%
96799-03-0
C7H7N3S
5 g
5-amino-3-2-thienyl pyrazole, 3-thiophen-2-yl-1h-pyrazol-5-amine, 5-thien-2-yl-1h-pyrazol-3-amine, 3-2-thienyl-1h-pyrazol-5-amine, 5-2-thienyl-1h-pyrazol-3-amine, 5-thiophen-2-yl-2h-pyrazol-3-ylamine, 5-thiophen-2-yl-1h-pyrazol-3-amine, gnf-pf-1850, 5-amino-3-thien-2-yl pyrazole, 5-amino-3-2-thienyl-1h-pyrazole
NC1=NNC(=C1)C1=CC=CS1
165.21
165.21

Chemical Identifiers

96799-03-0
165.21
TXSOLYKLZBJHFF-UHFFFAOYSA-N
523184
NC1=NNC(=C1)C1=CC=CS1
C7H7N3S
MFCD00051815
5-amino-3-2-thienyl pyrazole, 3-thiophen-2-yl-1h-pyrazol-5-amine, 5-thien-2-yl-1h-pyrazol-3-amine, 3-2-thienyl-1h-pyrazol-5-amine, 5-2-thienyl-1h-pyrazol-3-amine, 5-thiophen-2-yl-2h-pyrazol-3-ylamine, 5-thiophen-2-yl-1h-pyrazol-3-amine, gnf-pf-1850, 5-amino-3-thien-2-yl pyrazole, 5-amino-3-2-thienyl-1h-pyrazole
5-thiophen-2-yl-1H-pyrazol-3-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H302-H315-H318-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only