missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-2-nitrobenzotrifluoride 98.0+%, TCI America™
$34.66 - $115.51
Chemical Identifiers
| CAS | 393-11-3 |
|---|---|
| Molecular Formula | C7H5F3N2O2 |
| Molecular Weight (g/mol) | 206.12 |
| MDL Number | MFCD00014717 |
| InChI Key | UTKUVRNVYFTEHF-UHFFFAOYSA-N |
| Synonym | 4-nitro-3-trifluoromethyl aniline, 5-amino-2-nitrobenzotrifluoride, 3-amino-6-nitrobenzotrifluoride, benzenamine, 4-nitro-3-trifluoromethyl, 3-trifluoromethyl-4-nitroaniline, 2-nitro-5-amino benzotrifluoride, 3-trifluoromethyl-4-nitroanilide, flu-1, 3-amino-6-nitro benzotrifluoride |
| PubChem CID | 94955 |
| IUPAC Name | 4-nitro-3-(trifluoromethyl)aniline |
| SMILES | NC1=CC=C(C(=C1)C(F)(F)F)[N+]([O-])=O |
Chemical Identifiers
| 393-11-3 | |
| 206.12 | |
| UTKUVRNVYFTEHF-UHFFFAOYSA-N | |
| 94955 | |
| NC1=CC=C(C(=C1)C(F)(F)F)[N+]([O-])=O |
| C7H5F3N2O2 | |
| MFCD00014717 | |
| 4-nitro-3-trifluoromethyl aniline, 5-amino-2-nitrobenzotrifluoride, 3-amino-6-nitrobenzotrifluoride, benzenamine, 4-nitro-3-trifluoromethyl, 3-trifluoromethyl-4-nitroaniline, 2-nitro-5-amino benzotrifluoride, 3-trifluoromethyl-4-nitroanilide, flu-1, 3-amino-6-nitro benzotrifluoride | |
| 4-nitro-3-(trifluoromethyl)aniline |
Specifications
| 393-11-3 | |
| C7H5F3N2O2 | |
| 5 g | |
| UTKUVRNVYFTEHF-UHFFFAOYSA-N | |
| 4-nitro-3-(trifluoromethyl)aniline | |
| 94955 | |
| ≥98.0% (GC) | |
| 5-Amino-2-nitrobenzotrifluoride |
| 127°C | |
| MFCD00014717 | |
| 4-nitro-3-trifluoromethyl aniline, 5-amino-2-nitrobenzotrifluoride, 3-amino-6-nitrobenzotrifluoride, benzenamine, 4-nitro-3-trifluoromethyl, 3-trifluoromethyl-4-nitroaniline, 2-nitro-5-amino benzotrifluoride, 3-trifluoromethyl-4-nitroanilide, flu-1, 3-amino-6-nitro benzotrifluoride | |
| NC1=CC=C(C(=C1)C(F)(F)F)[N+]([O-])=O | |
| 206.12 | |
| 206.12 | |
| Crystalline Powder |
Safety and Handling
TSCA : No