missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-nitrobenzotrifluoride 98.0+%, TCI America™

Catalog No. A03775G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A03775G 5 g
A037725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A03775G Supplier TCI America Supplier No. A03775G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 393-11-3
Molecular Formula C7H5F3N2O2
Molecular Weight (g/mol) 206.12
MDL Number MFCD00014717
InChI Key UTKUVRNVYFTEHF-UHFFFAOYSA-N
Synonym 4-nitro-3-trifluoromethyl aniline, 5-amino-2-nitrobenzotrifluoride, 3-amino-6-nitrobenzotrifluoride, benzenamine, 4-nitro-3-trifluoromethyl, 3-trifluoromethyl-4-nitroaniline, 2-nitro-5-amino benzotrifluoride, 3-trifluoromethyl-4-nitroanilide, flu-1, 3-amino-6-nitro benzotrifluoride
PubChem CID 94955
IUPAC Name 4-nitro-3-(trifluoromethyl)aniline
SMILES NC1=CC=C(C(=C1)C(F)(F)F)[N+]([O-])=O

Specifications

Melting Point 127°C
Quantity 5 g
Formula Weight 206.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2-nitrobenzotrifluoride