missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-2-nitrobenzoic Acid 98.0+%, TCI America™
$182.89 - $641.08
Chemical Identifiers
| CAS | 13280-60-9 |
|---|---|
| Molecular Formula | C7H6N2O4 |
| Molecular Weight (g/mol) | 182.135 |
| MDL Number | MFCD00035748 |
| InChI Key | KZZWQCKYLNIOBT-UHFFFAOYSA-N |
| Synonym | 5-amino-2-nitrobenzoic acid, 2-nitro-5-aminobenzoic acid, benzoic acid, 5-amino-2-nitro, 3-carboxy-4-nitroaniline, 5-amino-2-nitro-benzoic acid, pubchem4983, acmc-1bxcb, intermediates-zcf02020, 5-amino-2-nitrobenzoicacid, 6-nitro-m-anthranilic acid |
| PubChem CID | 83298 |
| SMILES | C1=CC(=C(C=C1N)C(=O)O)[N+](=O)[O-] |
Chemical Identifiers
| 13280-60-9 | |
| 182.135 | |
| KZZWQCKYLNIOBT-UHFFFAOYSA-N | |
| 83298 |
| C7H6N2O4 | |
| MFCD00035748 | |
| 5-amino-2-nitrobenzoic acid, 2-nitro-5-aminobenzoic acid, benzoic acid, 5-amino-2-nitro, 3-carboxy-4-nitroaniline, 5-amino-2-nitro-benzoic acid, pubchem4983, acmc-1bxcb, intermediates-zcf02020, 5-amino-2-nitrobenzoicacid, 6-nitro-m-anthranilic acid | |
| C1=CC(=C(C=C1N)C(=O)O)[N+](=O)[O-] |
Specifications
| 13280-60-9 | |
| MFCD00035748 | |
| 5-amino-2-nitrobenzoic acid, 2-nitro-5-aminobenzoic acid, benzoic acid, 5-amino-2-nitro, 3-carboxy-4-nitroaniline, 5-amino-2-nitro-benzoic acid, pubchem4983, acmc-1bxcb, intermediates-zcf02020, 5-amino-2-nitrobenzoicacid, 6-nitro-m-anthranilic acid | |
| C1=CC(=C(C=C1N)C(=O)O)[N+](=O)[O-] | |
| 83298 | |
| ≥98.0% (T) | |
| 5-Amino-2-nitrobenzoic Acid |
| C7H6N2O4 | |
| 5 g | |
| KZZWQCKYLNIOBT-UHFFFAOYSA-N | |
| 182.135 | |
| 182.14 | |
| Crystalline Powder |
Safety and Handling
TSCA : No