missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-nitrobenzoic Acid 98.0+%, TCI America™

Catalog No. A08975G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A08975G 5 g
A089725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A08975G Supplier TCI America Supplier No. A08975G

Chemical Identifiers

CAS 13280-60-9
Molecular Formula C7H6N2O4
Molecular Weight (g/mol) 182.135
MDL Number MFCD00035748
InChI Key KZZWQCKYLNIOBT-UHFFFAOYSA-N
Synonym 5-amino-2-nitrobenzoic acid, 2-nitro-5-aminobenzoic acid, benzoic acid, 5-amino-2-nitro, 3-carboxy-4-nitroaniline, 5-amino-2-nitro-benzoic acid, pubchem4983, acmc-1bxcb, intermediates-zcf02020, 5-amino-2-nitrobenzoicacid, 6-nitro-m-anthranilic acid
PubChem CID 83298
SMILES C1=CC(=C(C=C1N)C(=O)O)[N+](=O)[O-]

Specifications

Quantity 5 g
Formula Weight 182.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2-nitrobenzoic Acid