missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-nitrobenzoic acid, 95%

Catalog No. AAL1622506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1622506 5 g
AAL1622514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1622506 Supplier Thermo Scientific Chemicals Supplier No. L1622506

CAS: 13280-60-9 | C7H6N2O4 | 182.135 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13280-60-9
Molecular Formula C7H6N2O4
Molecular Weight (g/mol) 182.135
MDL Number MFCD00035748
InChI Key KZZWQCKYLNIOBT-UHFFFAOYSA-N
Synonym 5-amino-2-nitrobenzoic acid, 2-nitro-5-aminobenzoic acid, benzoic acid, 5-amino-2-nitro, 3-carboxy-4-nitroaniline, 5-amino-2-nitro-benzoic acid, pubchem4983, acmc-1bxcb, intermediates-zcf02020, 5-amino-2-nitrobenzoicacid, 6-nitro-m-anthranilic acid
PubChem CID 83298
SMILES C1=CC(=C(C=C1N)C(=O)O)[N+](=O)[O-]

Specifications

Melting Point ∽240°C (decomposition)
Quantity 5 g
Beilstein 2107512
Formula Weight 182.14
Percent Purity 95%
Chemical Name or Material 5-Amino-2-nitrobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 236-283-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only