missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-naphthol 97.0+%, TCI America™

Catalog No. A03595G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A03595G 5 g
A035925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A03595G Supplier TCI America Supplier No. A03595G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 86-97-5
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.188
MDL Number MFCD00035729
InChI Key FSBRKZMSECKELY-UHFFFAOYSA-N
PubChem CID 6865
IUPAC Name 5-aminonaphthalen-2-ol
SMILES C1=CC2=C(C=CC(=C2)O)C(=C1)N

Specifications

Melting Point 183°C
Quantity 5 g
Formula Weight 159.19
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2-naphthol
EINECSNumber (4)-0363
RTECSNumber QL3330000
TSCA Yes