missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-2-naphthalenesulfonic Acid 97.0+%, TCI America™
Supplier: TCI America A0346500G
Specifications
| 5-Amino-2-naphthalenesulfonic Acid | |
| Gray-Red | |
| MFCD00004030 | |
| 5-amino-2-naphthalenesulfonic acid, 1,6-cleve's acid, 1-aminonaphthalene-6-sulfonic acid, 2-naphthalenesulfonic acid, 5-amino, 1-naphthylamine-6-sulfonic acid, kyselina cleve, cleve's acid-1,6, 1-amino-6-sulfonaphthalene, 5-naphthylamine-2-sulfonic acid, cleve's beta-acid | |
| C1=CC2=C(C=CC(=C2)S(=O)(=O)O)C(=C1)N | |
| 223.246 | |
| CHEBI:44188 | |
| ≥97.0% (T) |
| 119-79-9 | |
| C10H9NO3S | |
| 500 g | |
| UWPJYQYRSWYIGZ-UHFFFAOYSA-N | |
| 5-aminonaphthalene-2-sulfonic acid | |
| 8408 | |
| 223.25 | |
| Crystalline Powder |
Chemical Identifiers
| 119-79-9 | |
| 223.246 | |
| UWPJYQYRSWYIGZ-UHFFFAOYSA-N | |
| 8408 | |
| 5-aminonaphthalene-2-sulfonic acid |
| C10H9NO3S | |
| MFCD00004030 | |
| 5-amino-2-naphthalenesulfonic acid, 1,6-cleve's acid, 1-aminonaphthalene-6-sulfonic acid, 2-naphthalenesulfonic acid, 5-amino, 1-naphthylamine-6-sulfonic acid, kyselina cleve, cleve's acid-1,6, 1-amino-6-sulfonaphthalene, 5-naphthylamine-2-sulfonic acid, cleve's beta-acid | |
| CHEBI:44188 | |
| C1=CC2=C(C=CC(=C2)S(=O)(=O)O)C(=C1)N |
Safety and Handling
EINECSNumber : (4)-0491
RTECSNumber : QK1285000
TSCA : Yes