missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-2-mercaptobenzimidazole 97.0+%, TCI America™
$46.60 - $141.67
Chemical Identifiers
| CAS | 2818-66-8 |
|---|---|
| Molecular Formula | C7H7N3S |
| Molecular Weight (g/mol) | 165.21 |
| MDL Number | MFCD00022671 |
| InChI Key | BXDMTLVCACMNJO-UHFFFAOYSA-N |
| Synonym | 5-amino-2-mercaptobenzimidazole, 5-amino-2-benzimidazolethiol, 5-amino-1h-benzo d imidazole-2-thiol, 5-amino-1h-benzimidazole-2-thiol, 5-aminobenzimidazole-2-thiol, 2h-benzimidazole-2-thione, 5-amino-1,3-dihydro, 2-benzimidazolethiol, 5-amino, 5-amino-1h-1,3-benzodiazole-2-thiol, 2-mercapto-5-amino benzimidazole, 5-amino-3h-1,3-benzodiazole-2-thiol |
| PubChem CID | 2734002 |
| IUPAC Name | 5-amino-2,3-dihydro-1H-1,3-benzodiazole-2-thione |
| SMILES | NC1=CC=C2NC(=S)NC2=C1 |
Chemical Identifiers
| 2818-66-8 | |
| 165.21 | |
| BXDMTLVCACMNJO-UHFFFAOYSA-N | |
| 2734002 | |
| NC1=CC=C2NC(=S)NC2=C1 |
| C7H7N3S | |
| MFCD00022671 | |
| 5-amino-2-mercaptobenzimidazole, 5-amino-2-benzimidazolethiol, 5-amino-1h-benzo d imidazole-2-thiol, 5-amino-1h-benzimidazole-2-thiol, 5-aminobenzimidazole-2-thiol, 2h-benzimidazole-2-thione, 5-amino-1,3-dihydro, 2-benzimidazolethiol, 5-amino, 5-amino-1h-1,3-benzodiazole-2-thiol, 2-mercapto-5-amino benzimidazole, 5-amino-3h-1,3-benzodiazole-2-thiol | |
| 5-amino-2,3-dihydro-1H-1,3-benzodiazole-2-thione |
Specifications
| 2818-66-8 | |
| C7H7N3S | |
| 5 g | |
| BXDMTLVCACMNJO-UHFFFAOYSA-N | |
| 5-amino-2,3-dihydro-1H-1,3-benzodiazole-2-thione | |
| 2734002 | |
| ≥97.0% (T) | |
| 5-Amino-2-mercaptobenzimidazole |
| 237°C | |
| MFCD00022671 | |
| 5-amino-2-mercaptobenzimidazole, 5-amino-2-benzimidazolethiol, 5-amino-1h-benzo d imidazole-2-thiol, 5-amino-1h-benzimidazole-2-thiol, 5-aminobenzimidazole-2-thiol, 2h-benzimidazole-2-thione, 5-amino-1,3-dihydro, 2-benzimidazolethiol, 5-amino, 5-amino-1h-1,3-benzodiazole-2-thiol, 2-mercapto-5-amino benzimidazole, 5-amino-3h-1,3-benzodiazole-2-thiol | |
| NC1=CC=C2NC(=S)NC2=C1 | |
| 165.21 | |
| 165.21 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes