accessibility menu, dialog, popup

UserName

NH2HNSHN

CAS RN 2818-66-8

IUPAC Name: 5-amino-2,3-dihydro-1H-1,3-benzodiazole-2-thione
Synonyms: 5-amino-1,3-dihydro-1,3-benzodiazole-2-thione
Molecular Weight (g/mol): 165.21
Molecular Formula: C7H7N3S
InChi Key: BXDMTLVCACMNJO-UHFFFAOYSA-N
SMILES: NC1=CC=C2NC(=S)NC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results