missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-hydroxypyridine, 97%

Catalog No. p-7030332
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2796103 1 g
AAH2796106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2796103 Supplier Thermo Scientific Chemicals Supplier No. H2796103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 33630-94-3 | C5H6N2O | 110.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33630-94-3
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD03427652
InChI Key CHYPHWOOCGECMN-UHFFFAOYSA-N
Synonym 5-amino-2-hydroxypyridine, 5-aminopyridin-2-ol, 5-aminopyridin-2 1h-one, 3-amino-6-hydroxypyridine, 5-amino-2-pyridone, 5-amino-2-pyridinol, 2-hydroxy-5-aminopyridine, 2 1h-pyridinone, 5-amino, 5-amino-2 1h-pyridone
PubChem CID 818256
IUPAC Name 5-amino-1H-pyridin-2-one
SMILES NC1=CCC(=O)N=C1

Specifications

Melting Point ∽180°C (decomposition)
Quantity 1 g
Formula Weight 110.12
Percent Purity 97%
Chemical Name or Material 5-Amino-2-hydroxypyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only