accessibility menu, dialog, popup

UserName

CAS RN 33630-94-3

IUPAC Name: 5-amino-2,3-dihydropyridin-2-one
Synonyms: 5-amino-3H-pyridin-2-one
Molecular Weight (g/mol): 110.12
Molecular Formula: C5H6N2O
InChi Key: CHYPHWOOCGECMN-UHFFFAOYSA-N
SMILES: NC1=CCC(=O)N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results