missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-2-fluorobenzotrifluoride 98.0+%, TCI America™
$50.33 - $139.81
Chemical Identifiers
| CAS | 2357-47-3 |
|---|---|
| Molecular Formula | C7H5F4N |
| Molecular Weight (g/mol) | 179.118 |
| MDL Number | MFCD00007834 |
| InChI Key | PGFQDLOMDIBAPY-UHFFFAOYSA-N |
| Synonym | 4-fluoro-3-trifluoromethyl aniline, 5-amino-2-fluorobenzotrifluoride, alpha,alpha,alpha,4-tetrafluoro-m-toluidine, 3-trifluoromethyl-4-fluoroaniline, 4-fluoro-3-trifluoromethyl benzenamine, benzenamine, 4-fluoro-3-trifluoromethyl, unii-v5584x2ctl, 2-fluoro-5-aminobenzotrifluoride, 5-amino-2-fluoro trifluoromethyl benzene, 4-fluoro-3-trifluoromethyl phenylamine |
| PubChem CID | 75377 |
| IUPAC Name | 4-fluoro-3-(trifluoromethyl)aniline |
| SMILES | C1=CC(=C(C=C1N)C(F)(F)F)F |
Chemical Identifiers
| 2357-47-3 | |
| 179.118 | |
| PGFQDLOMDIBAPY-UHFFFAOYSA-N | |
| 75377 | |
| C1=CC(=C(C=C1N)C(F)(F)F)F |
| C7H5F4N | |
| MFCD00007834 | |
| 4-fluoro-3-trifluoromethyl aniline, 5-amino-2-fluorobenzotrifluoride, alpha,alpha,alpha,4-tetrafluoro-m-toluidine, 3-trifluoromethyl-4-fluoroaniline, 4-fluoro-3-trifluoromethyl benzenamine, benzenamine, 4-fluoro-3-trifluoromethyl, unii-v5584x2ctl, 2-fluoro-5-aminobenzotrifluoride, 5-amino-2-fluoro trifluoromethyl benzene, 4-fluoro-3-trifluoromethyl phenylamine | |
| 4-fluoro-3-(trifluoromethyl)aniline |
Specifications
| 2357-47-3 | |
| 208°C | |
| MFCD00007834 | |
| 4-fluoro-3-trifluoromethyl aniline, 5-amino-2-fluorobenzotrifluoride, alpha,alpha,alpha,4-tetrafluoro-m-toluidine, 3-trifluoromethyl-4-fluoroaniline, 4-fluoro-3-trifluoromethyl benzenamine, benzenamine, 4-fluoro-3-trifluoromethyl, unii-v5584x2ctl, 2-fluoro-5-aminobenzotrifluoride, 5-amino-2-fluoro trifluoromethyl benzene, 4-fluoro-3-trifluoromethyl phenylamine | |
| C1=CC(=C(C=C1N)C(F)(F)F)F | |
| 179.118 | |
| 179.12 | |
| Liquid |
| Yellow | |
| C7H5F4N | |
| 5 g | |
| PGFQDLOMDIBAPY-UHFFFAOYSA-N | |
| 4-fluoro-3-(trifluoromethyl)aniline | |
| 75377 | |
| ≥98.0% (GC) | |
| 5-Amino-2-fluorobenzotrifluoride |
Safety and Handling
TSCA : No