missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-2-cyanobenzotrifluoride 98.0+%, TCI America™
$53.90 - $130.92
Chemical Identifiers
| CAS | 654-70-6 |
|---|---|
| Molecular Formula | C8H5F3N2 |
| Molecular Weight (g/mol) | 186.137 |
| MDL Number | MFCD00042155 |
| InChI Key | PMDYLCUKSLBUHO-UHFFFAOYSA-N |
| Synonym | 4-amino-2-trifluoromethyl benzonitrile, 5-amino-2-cyanobenzotrifluoride, 4-cyano-3-trifluoromethylaniline, 2-cyano-5-aminobenzotrifluoride, 4-cyano-3-trifluoromethyl aniline, benzonitrile, 4-amino-2-trifluoromethyl, unii-l47d9xhc08, 4-amino 2-trifluoromethyl benzonitrile, 2-trifluoromethyl-4-aminobenzonitrile, 4-amino-2-trifluoromethyl benzenecarbonitrile |
| PubChem CID | 522170 |
| IUPAC Name | 4-amino-2-(trifluoromethyl)benzonitrile |
| SMILES | C1=CC(=C(C=C1N)C(F)(F)F)C#N |
Chemical Identifiers
| 654-70-6 | |
| 186.137 | |
| PMDYLCUKSLBUHO-UHFFFAOYSA-N | |
| 522170 | |
| C1=CC(=C(C=C1N)C(F)(F)F)C#N |
| C8H5F3N2 | |
| MFCD00042155 | |
| 4-amino-2-trifluoromethyl benzonitrile, 5-amino-2-cyanobenzotrifluoride, 4-cyano-3-trifluoromethylaniline, 2-cyano-5-aminobenzotrifluoride, 4-cyano-3-trifluoromethyl aniline, benzonitrile, 4-amino-2-trifluoromethyl, unii-l47d9xhc08, 4-amino 2-trifluoromethyl benzonitrile, 2-trifluoromethyl-4-aminobenzonitrile, 4-amino-2-trifluoromethyl benzenecarbonitrile | |
| 4-amino-2-(trifluoromethyl)benzonitrile |
Specifications
| 654-70-6 | |
| C8H5F3N2 | |
| 1 g | |
| 4-amino-2-trifluoromethyl benzonitrile, 5-amino-2-cyanobenzotrifluoride, 4-cyano-3-trifluoromethylaniline, 2-cyano-5-aminobenzotrifluoride, 4-cyano-3-trifluoromethyl aniline, benzonitrile, 4-amino-2-trifluoromethyl, unii-l47d9xhc08, 4-amino 2-trifluoromethyl benzonitrile, 2-trifluoromethyl-4-aminobenzonitrile, 4-amino-2-trifluoromethyl benzenecarbonitrile | |
| C1=CC(=C(C=C1N)C(F)(F)F)C#N | |
| 186.137 | |
| 186.14 | |
| Crystalline Powder |
| 142°C | |
| MFCD00042155 | |
| 3439 | |
| PMDYLCUKSLBUHO-UHFFFAOYSA-N | |
| 4-amino-2-(trifluoromethyl)benzonitrile | |
| 522170 | |
| ≥98.0% (GC) | |
| 5-Amino-2-cyanobenzotrifluoride |
Safety and Handling
TSCA : No