missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-cyanobenzotrifluoride 98.0+%, TCI America™

Catalog No. A12131G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12131G 1 g
A12135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A12131G Supplier TCI America Supplier No. A12131G

Chemical Identifiers

CAS 654-70-6
Molecular Formula C8H5F3N2
Molecular Weight (g/mol) 186.137
MDL Number MFCD00042155
InChI Key PMDYLCUKSLBUHO-UHFFFAOYSA-N
Synonym 4-amino-2-trifluoromethyl benzonitrile, 5-amino-2-cyanobenzotrifluoride, 4-cyano-3-trifluoromethylaniline, 2-cyano-5-aminobenzotrifluoride, 4-cyano-3-trifluoromethyl aniline, benzonitrile, 4-amino-2-trifluoromethyl, unii-l47d9xhc08, 4-amino 2-trifluoromethyl benzonitrile, 2-trifluoromethyl-4-aminobenzonitrile, 4-amino-2-trifluoromethyl benzenecarbonitrile
PubChem CID 522170
IUPAC Name 4-amino-2-(trifluoromethyl)benzonitrile
SMILES C1=CC(=C(C=C1N)C(F)(F)F)C#N

Specifications

Melting Point 142°C
Quantity 1 g
UN Number 3439
Formula Weight 186.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2-cyanobenzotrifluoride