missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-2-chlorotoluene-4-sulfonic Acid 98.0+%, TCI America™
$50.33 - $96.94
Chemical Identifiers
| CAS | 88-53-9 |
|---|---|
| Molecular Formula | C7H8ClNO3S |
| Molecular Weight (g/mol) | 221.655 |
| MDL Number | MFCD00043924 |
| InChI Key | VYZCFAPUHSSYCC-UHFFFAOYSA-N |
| Synonym | 4-Chloro-5-methylaniline-2-sulfonic Acid, 4-Chloro-m-toluidine-6-sulfonic Acid, 2-Amino-5-chloro-4-methylbenzenesulfonic Acid |
| PubChem CID | 6936 |
| IUPAC Name | 2-amino-5-chloro-4-methylbenzenesulfonic acid |
| SMILES | CC1=CC(=C(C=C1Cl)S(=O)(=O)O)N |
Chemical Identifiers
| 88-53-9 | |
| 221.655 | |
| VYZCFAPUHSSYCC-UHFFFAOYSA-N | |
| 6936 | |
| CC1=CC(=C(C=C1Cl)S(=O)(=O)O)N |
| C7H8ClNO3S | |
| MFCD00043924 | |
| 4-Chloro-5-methylaniline-2-sulfonic Acid, 4-Chloro-m-toluidine-6-sulfonic Acid, 2-Amino-5-chloro-4-methylbenzenesulfonic Acid | |
| 2-amino-5-chloro-4-methylbenzenesulfonic acid |
Specifications
| 88-53-9 | |
| MFCD00043924 | |
| 4-Chloro-5-methylaniline-2-sulfonic Acid, 4-Chloro-m-toluidine-6-sulfonic Acid, 2-Amino-5-chloro-4-methylbenzenesulfonic Acid | |
| CC1=CC(=C(C=C1Cl)S(=O)(=O)O)N | |
| 221.655 | |
| 221.66 | |
| Crystalline Powder |
| C7H8ClNO3S | |
| 25 g | |
| VYZCFAPUHSSYCC-UHFFFAOYSA-N | |
| 2-amino-5-chloro-4-methylbenzenesulfonic acid | |
| 6936 | |
| ≥98.0% (HPLC,T) | |
| 5-Amino-2-chlorotoluene-4-sulfonic Acid |
Safety and Handling
EINECSNumber : (3)-2024
RTECSNumber : DB4807000
TSCA : Yes