missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-chloropyridine 98.0+%, TCI America™

Catalog No. p-7117272
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
A11551G 1 g
A11555G 5 g
2 options

Catalog No. A11551G

Supplier: TCI America A11551G

Only null left

Chemical Identifiers

CAS 5350-93-6
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.56
MDL Number MFCD00006243
InChI Key QAJYCQZQLVENRZ-UHFFFAOYSA-N
Synonym 5-amino-2-chloropyridine, 2-chloro-5-aminopyridine, 3-amino-6-chloropyridine, 3-pyridinamine, 6-chloro, 6-chloro-pyridin-3-ylamine, 6-chloro-3-pyridylamine, 2-chloro-5-amino pyridine, 6-chloro-3-pyridinamine, 6-chloropyridin-3-ylamine, 6-chlorpyridin-3-amin
PubChem CID 79305
IUPAC Name 6-chloropyridin-3-amine
SMILES NC1=CC=C(Cl)N=C1

Specifications

Melting Point 83°C
Quantity 1 g
Formula Weight 128.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2-chloropyridine