accessibility menu, dialog, popup

UserName

CAS RN 5350-93-6

NH2ClN

CAS RN 5350-93-6

IUPAC Name: 6-chloropyridin-3-amine
Synonyms: 6-chloropyridin-3-amine
Molecular Weight (g/mol): 128.56
Molecular Formula: C5H5ClN2
InChi Key: QAJYCQZQLVENRZ-UHFFFAOYSA-N
SMILES: NC1=CC=C(Cl)N=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results