missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-chlorobenzotrifluoride 98.0+%, TCI America™

Catalog No. p-7124685
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C09225G 5 g
C092225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C09225G Supplier TCI America Supplier No. C09225G
Only null left

Chemical Identifiers

CAS 320-51-4
Molecular Formula C7H5ClF3N
Molecular Weight (g/mol) 195.569
MDL Number MFCD00007846
InChI Key ASPDJZINBYYZRU-UHFFFAOYSA-N
Synonym 4-chloro-3-trifluoromethyl aniline, 5-amino-2-chlorobenzotrifluoride, 3-trifluoromethyl-4-chloroaniline, 4-chloro-alpha,alpha,alpha-trifluoro-m-toluidine, 2-chloro-5-aminobenzotrifluoride, acbt, benzenamine, 4-chloro-3-trifluoromethyl, 2-chloro-5-aminebenzotrifluoride, 4-chloro-3-trifluoromethyl-phenylamine, 4-chloro-3-trifluoromethyl benzenamine
PubChem CID 67574
ChEBI CHEBI:59245
IUPAC Name 4-chloro-3-(trifluoromethyl)aniline
SMILES C1=CC(=C(C=C1N)C(F)(F)F)Cl

Specifications

Quantity 5 g
UN Number 2811
Formula Weight 195.57
Percent Purity ≥98.0% (GC,T)
Physical Form Crystals
Chemical Name or Material 5-Amino-2-chlorobenzotrifluoride
EINECSNumber (3)-0266
TSCA Yes