missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-chlorobenzotrifluoride, 99%

Catalog No. AC103371000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC103371000 100 g Glass bottle
AC103370050 5 g Glass bottle
AC103370250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC103371000 Supplier Thermo Scientific Chemicals Supplier No. 103371000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 320-51-4 | C7H5ClF3N | 195.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 320-51-4
Molecular Formula C7H5ClF3N
Molecular Weight (g/mol) 195.57
MDL Number MFCD00007846
InChI Key ASPDJZINBYYZRU-UHFFFAOYSA-N
Synonym 4-chloro-3-trifluoromethyl aniline, 5-amino-2-chlorobenzotrifluoride, 3-trifluoromethyl-4-chloroaniline, 4-chloro-alpha,alpha,alpha-trifluoro-m-toluidine, 2-chloro-5-aminobenzotrifluoride, acbt, benzenamine, 4-chloro-3-trifluoromethyl, 2-chloro-5-aminebenzotrifluoride, 4-chloro-3-trifluoromethyl-phenylamine, 4-chloro-3-trifluoromethyl benzenamine
PubChem CID 67574
ChEBI CHEBI:59245
IUPAC Name 4-chloro-3-(trifluoromethyl)aniline
SMILES C1=CC(=C(C=C1N)C(F)(F)F)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 34.0°C to 38.0°C
Color Orange or Pink to White
Boiling Point 85.0°C (3.0 mmHg)
Flash Point 134°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H3(Cl)CF3
Quantity 100 g
Solubility Information Solubility in water: negligible
Formula Weight 195.57
Percent Purity 99%
Physical Form Crystalline Powder or Low Melting Solid
Chemical Name or Material 5-Amino-2-chlorobenzotrifluoride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medi
EINECSNumber 206-277-6
TSCA TSCA

RUO – Research Use Only