missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-chlorobenzoic Acid 98.0+%, TCI America™

Catalog No. A191325G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A191325G 25 g
A1913250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A191325G Supplier TCI America Supplier No. A191325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 89-54-3
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007845
InChI Key GVCFFVPEOLCYNN-UHFFFAOYSA-N
Synonym 2-chloro-5-aminobenzoic acid, benzoic acid, 5-amino-2-chloro, 3-amino-6-chlorobenzoic acid, 5-amino-2-chloro-benzoic acid, 5-amino-2-chlorobenzoicacid, 3-carboxy-4-chloroaniline, 5-amino-2-chloro benzoic acid, zlchem 462, pubchem4654, acmc-209r1l
PubChem CID 37879
IUPAC Name 5-amino-2-chlorobenzoic acid
SMILES NC1=CC=C(Cl)C(=C1)C(O)=O

Specifications

Melting Point 188°C
Color White-Yellow
Quantity 25 g
Formula Weight 171.58
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2-chlorobenzoic Acid
EINECSNumber (3)-1480
TSCA Yes