missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-bromo-4-methylpyridine, 98%

CAS: 156118-16-0 | C6H7BrN2 | 187.04 g/mol

$110.86 - $467.81

Chemical Identifiers

CAS 156118-16-0
Molecular Formula C6H7BrN2
Molecular Weight (g/mol) 187.04
MDL Number MFCD08277280
InChI Key MVDBPMJQCXZKRB-UHFFFAOYSA-N
Synonym 3-amino-6-bromo-4-methylpyridine, 2-bromo-4-methyl-5-aminopyridine, 5-amino-2-bromo-4-picoline, 2-bromo-5-amino-4-picoline, 5-amino-2-bromo-4-methylpyridine, 6-bromo-4-methyl-pyridin-3-ylamine, 6-bromo-4-methyl-3-pyridinamine, 3-pyridinamine, 6-bromo-4-methyl, 6-bromo-4-methyl-3-pyridylamine, pubchem5960
PubChem CID 19926094
IUPAC Name 6-bromo-4-methylpyridin-3-amine
SMILES CC1=CC(=NC=C1N)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6695703
View Documents
Thermo Scientific Chemicals
H6695703
1 g
Each for $110.86
Only null left
 
AAH6695706
View Documents
Thermo Scientific Chemicals
H6695706
5 g
Each for $467.81
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

156118-16-0
187.04
MVDBPMJQCXZKRB-UHFFFAOYSA-N
19926094
CC1=CC(=NC=C1N)Br
C6H7BrN2
MFCD08277280
3-amino-6-bromo-4-methylpyridine, 2-bromo-4-methyl-5-aminopyridine, 5-amino-2-bromo-4-picoline, 2-bromo-5-amino-4-picoline, 5-amino-2-bromo-4-methylpyridine, 6-bromo-4-methyl-pyridin-3-ylamine, 6-bromo-4-methyl-3-pyridinamine, 3-pyridinamine, 6-bromo-4-methyl, 6-bromo-4-methyl-3-pyridylamine, pubchem5960
6-bromo-4-methylpyridin-3-amine

Specifications

156118-16-0
C6H7BrN2
1 g
10654630
MVDBPMJQCXZKRB-UHFFFAOYSA-N
6-bromo-4-methylpyridin-3-amine
19926094
98%
5-Amino-2-bromo-4-methylpyridine
Brown
MFCD08277280
UN2811
3-amino-6-bromo-4-methylpyridine, 2-bromo-4-methyl-5-aminopyridine, 5-amino-2-bromo-4-picoline, 2-bromo-5-amino-4-picoline, 5-amino-2-bromo-4-methylpyridine, 6-bromo-4-methyl-pyridin-3-ylamine, 6-bromo-4-methyl-3-pyridinamine, 3-pyridinamine, 6-bromo-4-methyl, 6-bromo-4-methyl-3-pyridylamine, pubchem5960
CC1=CC(=NC=C1N)Br
187.04
187.04
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only