CAS RN 156118-16-0
IUPAC Name:
6-bromo-4-methylpyridin-3-amine
Synonyms:
6-bromo-4-methylpyridin-3-amine
Molecular Weight (g/mol):
187.04
Molecular Formula:
C6H7BrN2
InChi Key:
MVDBPMJQCXZKRB-UHFFFAOYSA-N
SMILES:
CC1=CC(Br)=NC=C1N
1
–
1
of
1
results
1
Promotions Available
5-Amino-2-bromo-4-methylpyridine, 98%
5-Amino-2-bromo-4-methylpyridine, 98%
CAS: 156118-16-0 Molecular Formula: C6H7BrN2 Molecular Weight (g/mol): 187.04 MDL Number: MFCD08277280 InChI Key: MVDBPMJQCXZKRB-UHFFFAOYSA-N Synonym: 3-amino-6-bromo-4-methylpyridine,2-bromo-4-methyl-5-aminopyridine,5-amino-2-bromo-4-picoline,2-bromo-5-amino-4-picoline,5-amino-2-bromo-4-methylpyridine,6-bromo-4-methyl-pyridin-3-ylamine,6-bromo-4-methyl-3-pyridinamine,3-pyridinamine, 6-bromo-4-methyl,6-bromo-4-methyl-3-pyridylamine,pubchem5960 PubChem CID: 19926094 IUPAC Name: 6-bromo-4-methylpyridin-3-amine SMILES: CC1=CC(=NC=C1N)Br