missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-bromo-4-methylpyridine, 98%

Catalog No. AAH6695703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6695703 1 g
AAH6695706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6695703 Supplier Thermo Scientific Chemicals Supplier No. H6695703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 156118-16-0 | C6H7BrN2 | 187.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 156118-16-0
Molecular Formula C6H7BrN2
Molecular Weight (g/mol) 187.04
MDL Number MFCD08277280
InChI Key MVDBPMJQCXZKRB-UHFFFAOYSA-N
Synonym 3-amino-6-bromo-4-methylpyridine, 2-bromo-4-methyl-5-aminopyridine, 5-amino-2-bromo-4-picoline, 2-bromo-5-amino-4-picoline, 5-amino-2-bromo-4-methylpyridine, 6-bromo-4-methyl-pyridin-3-ylamine, 6-bromo-4-methyl-3-pyridinamine, 3-pyridinamine, 6-bromo-4-methyl, 6-bromo-4-methyl-3-pyridylamine, pubchem5960
PubChem CID 19926094
IUPAC Name 6-bromo-4-methylpyridin-3-amine
SMILES CC1=CC(=NC=C1N)Br

Specifications

Color Brown
Quantity 1 g
UN Number UN2811
Beilstein 10654630
Formula Weight 187.04
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Amino-2-bromo-4-methylpyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only