missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-2-bromo-3-methylpyridine 95.0+%, TCI America™
$42.36 - $117.44
Chemical Identifiers
| CAS | 38186-83-3 |
|---|---|
| Molecular Formula | C6H7BrN2 |
| Molecular Weight (g/mol) | 187.04 |
| MDL Number | MFCD03095085 |
| InChI Key | IEONGQZSAUAAKH-UHFFFAOYSA-N |
| Synonym | 5-amino-2-bromo-3-picoline, 5-amino-2-bromo-3-methylpyridine, 3-pyridinamine, 6-bromo-5-methyl, 6-bromo-5-methyl-3-pyridylamine, 6-bromo-5-methyl-3-aminopyridine, pubchem1129, ksc985c1p, 2-bromo-3-methyl-5-aminopyridine, 2-bromo-5-amino-3-methylpyridine, 6-bromo-5-methyl-pyridin-3-ylamine |
| PubChem CID | 12362446 |
| IUPAC Name | 6-bromo-5-methylpyridin-3-amine |
| SMILES | CC1=CC(N)=CN=C1Br |
Chemical Identifiers
| 38186-83-3 | |
| 187.04 | |
| IEONGQZSAUAAKH-UHFFFAOYSA-N | |
| 12362446 | |
| CC1=CC(N)=CN=C1Br |
| C6H7BrN2 | |
| MFCD03095085 | |
| 5-amino-2-bromo-3-picoline, 5-amino-2-bromo-3-methylpyridine, 3-pyridinamine, 6-bromo-5-methyl, 6-bromo-5-methyl-3-pyridylamine, 6-bromo-5-methyl-3-aminopyridine, pubchem1129, ksc985c1p, 2-bromo-3-methyl-5-aminopyridine, 2-bromo-5-amino-3-methylpyridine, 6-bromo-5-methyl-pyridin-3-ylamine | |
| 6-bromo-5-methylpyridin-3-amine |
Specifications
| 38186-83-3 | |
| C6H7BrN2 | |
| 1 g | |
| IEONGQZSAUAAKH-UHFFFAOYSA-N | |
| 6-bromo-5-methylpyridin-3-amine | |
| 12362446 | |
| ≥95.0% (GC,T) | |
| 5-Amino-2-bromo-3-methylpyridine |
| 100°C | |
| MFCD03095085 | |
| 5-amino-2-bromo-3-picoline, 5-amino-2-bromo-3-methylpyridine, 3-pyridinamine, 6-bromo-5-methyl, 6-bromo-5-methyl-3-pyridylamine, 6-bromo-5-methyl-3-aminopyridine, pubchem1129, ksc985c1p, 2-bromo-3-methyl-5-aminopyridine, 2-bromo-5-amino-3-methylpyridine, 6-bromo-5-methyl-pyridin-3-ylamine | |
| CC1=CC(N)=CN=C1Br | |
| 187.04 | |
| 187.04 | |
| Crystalline Powder |
Safety and Handling
TSCA : No