accessibility menu, dialog, popup

UserName

CH3NBrNH2

CAS RN 38186-83-3

IUPAC Name: 6-bromo-5-methylpyridin-3-amine
Synonyms: 6-bromo-5-methylpyridin-3-amine
Molecular Weight (g/mol): 187.04
Molecular Formula: C6H7BrN2
InChi Key: IEONGQZSAUAAKH-UHFFFAOYSA-N
SMILES: CC1=CC(N)=CN=C1Br
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)