missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-bromo-3-methylpyridine 95.0+%, TCI America™

Catalog No. A29751G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29751G 1 g
A29755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29751G Supplier TCI America Supplier No. A29751G

Chemical Identifiers

CAS 38186-83-3
Molecular Formula C6H7BrN2
Molecular Weight (g/mol) 187.04
MDL Number MFCD03095085
InChI Key IEONGQZSAUAAKH-UHFFFAOYSA-N
Synonym 5-amino-2-bromo-3-picoline, 5-amino-2-bromo-3-methylpyridine, 3-pyridinamine, 6-bromo-5-methyl, 6-bromo-5-methyl-3-pyridylamine, 6-bromo-5-methyl-3-aminopyridine, pubchem1129, ksc985c1p, 2-bromo-3-methyl-5-aminopyridine, 2-bromo-5-amino-3-methylpyridine, 6-bromo-5-methyl-pyridin-3-ylamine
PubChem CID 12362446
IUPAC Name 6-bromo-5-methylpyridin-3-amine
SMILES CC1=CC(N)=CN=C1Br

Specifications

Melting Point 100°C
Quantity 1 g
Formula Weight 187.04
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2-bromo-3-methylpyridine