missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-2-benzimidazolinone 98.0+%, TCI America™
$32.72 - $42.36
Chemical Identifiers
| CAS | 95-23-8 |
|---|---|
| Molecular Formula | C7H7N3O |
| Molecular Weight (g/mol) | 149.153 |
| MDL Number | MFCD00053555 |
| InChI Key | BCXSVFBDMPSKPT-UHFFFAOYSA-N |
| PubChem CID | 66765 |
| IUPAC Name | 5-amino-1,3-dihydrobenzimidazol-2-one |
| SMILES | C1=CC2=C(C=C1N)NC(=O)N2 |
Chemical Identifiers
| 95-23-8 | |
| 149.153 | |
| BCXSVFBDMPSKPT-UHFFFAOYSA-N | |
| 5-amino-1,3-dihydrobenzimidazol-2-one |
| C7H7N3O | |
| MFCD00053555 | |
| 66765 | |
| C1=CC2=C(C=C1N)NC(=O)N2 |
Specifications
| 95-23-8 | |
| C7H7N3O | |
| 5 g | |
| C1=CC2=C(C=C1N)NC(=O)N2 | |
| 149.153 | |
| 149.15 | |
| Crystalline Powder |
| Purple-White | |
| MFCD00053555 | |
| BCXSVFBDMPSKPT-UHFFFAOYSA-N | |
| 5-amino-1,3-dihydrobenzimidazol-2-one | |
| 66765 | |
| ≥98.0% (HPLC) | |
| 5-Amino-2-benzimidazolinone |
Safety and Handling
TSCA : Yes