missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2,4-dichloroanisole 98.0+%, TCI America™

Catalog No. p-7117721
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20855G 5 g
A208525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20855G Supplier TCI America Supplier No. A20855G
Only null left

Chemical Identifiers

CAS 98446-49-2
Molecular Formula C7H7Cl2NO
Molecular Weight (g/mol) 192.039
MDL Number MFCD00974410
InChI Key AJROJTARXSATEB-UHFFFAOYSA-N
Synonym 5-amino-2,4-dichloroanisole, 2,4-dichloro-5-methoxybenzenamine, 4,6-dichloro-m-anisidine, 2,4-dichloro-5-methoxy-phenylamine, benzenamine, 2,4-dichloro-5-methoxy, 2,4-dichloro-5-methoxyphenylamine, dichloromethoxyaniline, pubchem4393, acmc-209sae, 4,5-dichloro-m-anisidine
PubChem CID 1476636
IUPAC Name 2,4-dichloro-5-methoxyaniline
SMILES COC1=C(C=C(C(=C1)N)Cl)Cl

Specifications

Melting Point 51°C
Quantity 5 g
Formula Weight 192.04
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2,4-dichloroanisole