missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-2-(4-aminophenyl)benzimidazole 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 236°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 224.27 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 5-Amino-2-(4-aminophenyl)benzimidazole |
Safety and Handling
| RTECSNumber | DD5421200 |
| TSCA | Yes |