accessibility menu, dialog, popup

UserName

H2NHNNH2N

CAS RN 7621-86-5

IUPAC Name: 2-(4-aminophenyl)-1H-1,3-benzodiazol-6-amine
Synonyms: 2-(4-aminophenyl)-3H-1,3-benzodiazol-5-amine
Molecular Weight (g/mol): 224.27
Molecular Formula: C13H12N4
InChi Key: XAFOTXWPFVZQAZ-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C1=NC2=CC=C(N)C=C2N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results