missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-(4-aminophenyl)benzimidazole 98.0+%, TCI America™

Catalog No. A27591G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27591G 1 g
A27595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27591G Supplier TCI America Supplier No. A27591G
Only null left

Chemical Identifiers

CAS 7621-86-5
Molecular Formula C13H12N4
Molecular Weight (g/mol) 224.27
MDL Number MFCD00043996,MFCD00451475
InChI Key XAFOTXWPFVZQAZ-UHFFFAOYSA-N
PubChem CID 24260
IUPAC Name 2-(4-aminophenyl)-1H-1,3-benzodiazol-6-amine
SMILES NC1=CC=C(C=C1)C1=NC2=CC=C(N)C=C2N1

Specifications

Melting Point 236°C
Color White-Yellow
Quantity 1 g
Formula Weight 224.27
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2-(4-aminophenyl)benzimidazole
RTECSNumber DD5421200
TSCA Yes