missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-2,4,6-triiodoisophthaloyl Dichloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 231°C |
| Quantity | 5 g |
| UN Number | 3077 |
| Formula Weight | 595.72 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 5-Amino-2,4,6-triiodoisophthaloyl Dichloride |
Safety and Handling
| TSCA | No |