missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1H-tetrazole 98.0+%, TCI America™

Catalog No. p-7116896
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A042725G 25 g
A0427100G 100 g
A0427500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A042725G Supplier TCI America Supplier No. A042725G
Only null left

Chemical Identifiers

CAS 4418-61-5
Molecular Formula CH3N5
Molecular Weight (g/mol) 85.07
MDL Number MFCD00005248
InChI Key ULRPISSMEBPJLN-UHFFFAOYSA-N
Synonym 5-aminotetrazole, 5-amino-1h-tetrazole, 1h-tetrazol-5-amine, aminotetrazole, tetrazol-5-ylamine, 1h-tetrazole, 5-amino, 5-amino-1,2,3,4-tetrazole, 5-amino tetrazole, 1h-tetrazol-5-ylamine, 5-amino-1h-tetrazol
PubChem CID 20467
IUPAC Name 2H-tetrazol-5-amine
SMILES C1(=NNN=N1)N

Specifications

Melting Point 201°C
Color White
Quantity 25 g
UN Number 1325
Formula Weight 85.07
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-1H-tetrazole
EINECSNumber (5)-0627
RTECSNumber XF7465000
TSCA Yes