missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1H-benzotriazole, 96%

Catalog No. p-7044476
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0666003 1 g
AAL0666006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0666003 Supplier Thermo Scientific Chemicals Supplier No. L0666003
Only null left

CAS: 3325-11-9 | C6H6N4 | 134.14 g/mol

5-Amino-1H-benzotriazole is used as azole building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Amino-1H-benzotriazole is used as azole building block.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.

Chemical Identifiers

CAS 3325-11-9
Molecular Formula C6H6N4
Molecular Weight (g/mol) 134.14
MDL Number MFCD00047209
InChI Key XSFHICWNEBCMNN-UHFFFAOYSA-N
Synonym 5-aminobenzotriazole, 1h-1,2,3-benzotriazol-5-amine, 1h-benzotriazol-6-amine, 1h-benzotriazol-5-amine, 5-amino-1h-benzotriazole, 2h-benzo d 1,2,3 triazol-5-amine, 1h-benzo d 1,2,3 triazol-5-amine, benzotriazole-5-ylamine, 3h-1,2,3-benzotriazol-5-amine, 5-amino benzotriazole
PubChem CID 76844
IUPAC Name 2H-benzotriazol-5-amine
SMILES C1=CC2=NNN=C2C=C1N

Specifications

Melting Point 157°C to 162°C
Odor Odorless
Quantity 1 g
Beilstein 121262
Solubility Information Insoluble in water.
Formula Weight 134.14
Percent Purity 98%
Chemical Name or Material 5-Amino-1H-benzotriazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only