missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G-Biosciences 5-alpha Chemically Competent Cells
Description
It is T1 phage resistant and endA deficient for high-quality plasmid preparations. Chemically competent 5-alpha E. coli cells suitable for high efficiency transformation in a wide variety of applications.
Specifications
Specifications
| Product Type | Competent Cells |
| Sterility | Sterile |
| Species | E. coli |
| Format | Vial |
| For Use With (Application) | DNA Transformation |
| Quantity | 12 x 50 μL |