missing translation for 'onlineSavingsMsg'
Learn More

G-Biosciences 5-alpha Chemically Competent Cells

Catalog No. S000762
Encompass
Change view
Click to view available options
Quantity:
12 x 50 μL
6 x 100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000762 12 x 50 μL
S000759 6 x 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S000762 Supplier G-Biosciences Supplier No. 7862025
Only null left

5-alpha Chemically competent cells are a derivative of the popular DH5a.

It is T1 phage resistant and endA deficient for high-quality plasmid preparations. Chemically competent 5-alpha E. coli cells suitable for high efficiency transformation in a wide variety of applications.

Specifications

Product Type Competent Cells
Sterility Sterile
Species E. coli
Format Vial
For Use With (Application) DNA Transformation
Quantity 12 x 50 μL