missing translation for 'onlineSavingsMsg'
Learn More

5-Allyl-2-hydroxy-3-methoxybenzaldehyde, 98%

Catalog No. p-3212191
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC379360100 10 g Glass bottle
AC379360250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC379360100 Supplier Thermo Scientific Chemicals Supplier No. 379360100

CAS: 22934-51-6 | C11H12O3 | 192.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22934-51-6
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.21
MDL Number MFCD06408022
InChI Key FDHXEIOBIOVBEN-UHFFFAOYSA-N
Synonym 5-allyl-2-hydroxy-3-methoxybenzaldehyde, 5-allyl-3-methoxysalicylaldehyde, benzaldehyde, 2-hydroxy-3-methoxy-5-2-propenyl, benzaldehyde,2-hydroxy-3-methoxy-5-2-propen-1-yl, 2-hydroxy-3-methoxy-5-prop-2-en-1-yl benzaldehyde, formyl eugenol, acmc-209fzq, 2-hydroxy-3-methoxy-5-2-propenyl benzaldehyde, 3-methoxy-2-oxidanyl-5-prop-2-enyl-benzaldehyde
PubChem CID 1713887
IUPAC Name 2-hydroxy-3-methoxy-5-prop-2-enylbenzaldehyde
SMILES COC1=C(C(=CC(=C1)CC=C)C=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 48.0°C to 52.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 10 g
Solubility Information Solubility in water: insoluble
Formula Weight 192.21
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Allyl-2-hydroxy-3-methoxybenzaldehyde
Show More Show Less

RUO – Research Use Only