accessibility menu, dialog, popup

UserName

H3COCH2OHO

CAS RN 22934-51-6

IUPAC Name: 2-hydroxy-3-methoxy-5-(prop-2-en-1-yl)benzaldehyde
Synonyms: 2-hydroxy-3-methoxy-5-(prop-2-en-1-yl)benzaldehyde
Molecular Weight (g/mol): 192.21
Molecular Formula: C11H12O3
InChi Key: FDHXEIOBIOVBEN-UHFFFAOYSA-N
SMILES: COC1=CC(CC=C)=CC(C=O)=C1O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)